Learn More
TASP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16739-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes an endopeptidase that cleaves specific substrates following aspartate residues. The encoded protein undergoes posttranslational autoproteolytic processing to generate alpha and beta subunits, which reassemble into the active alpha2-beta2 heterotetramer. It is required to cleave MLL, a protein required for the maintece of HOX gene expression, and TFIIA, a basal transcription factor. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
| TASP1 | |
| Polyclonal | |
| Unconjugated | |
| TASP1 | |
| C20orf13, TASP1, Taspase 1, Threonine aspartase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 311468, 55617, 75812 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R1G1, Q9H6P5 | |
| Tasp1 | |
| TASP1 Fusion Protein Ag9918 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |