missing translation for 'onlineSavingsMsg'
Learn More

Tavaborole (>90%), TRC

Product Code. 30415109
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415109 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415109 Supplier Toronto Research Chemicals Supplier No. TRCT0097601G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tavaborole
CAS 174671-46-6
Molecular Formula C7 H6 B F O2
Quantity 1 g
Synonym 2,1-Benzoxaborole','5-fluoro-1,3-dihydro-1-hydroxy-, 5-Fluoro-1,3-dihydro-1-hydroxy-2,1-benzoxaborole, 5-Fluoro-1,3-dihydro-2,1-benzoxaborol-1-ol, AN 2690, AN 2729, Kerydin, Tavaborole
InChI Formula InChI=1S/C7H6BFO2/c9-6-1-2-7-5(3-6)4-11-8(7)10/h1-3,10H,4H2
SMILES OB1OCc2cc(F)ccc12
IUPAC Name 5-fluoro-1-hydroxy-3H-2,1-benzoxaborole
Molecular Weight (g/mol) 151.93
Formula Weight 152.0445
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.