missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAZ/WWTR1 Antibody (CL0371) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52926IR
This item is not returnable.
View return policy
Description
TAZ/WWTR1 Monoclonal antibody specifically detects TAZ/WWTR1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| TAZ/WWTR1 | |
| Monoclonal | |
| DyLight 755 | |
| FLJ27004, FLJ45718, TAZDKFZp586I1419, transcriptional co-activator with PDZ-binding motif, Transcriptional coactivator with PDZ-binding motif, WW domain containing transcription regulator 1, WW domain-containing transcription regulator protein 1 | |
| This antibody was developed using a recombinant protein derived from Q9GZV5, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Chromatin Research, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators | |
| 25937 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0371 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction