missing translation for 'onlineSavingsMsg'
Learn More

TBDMS-(3Beta)-3-[(Tetrahydro-2H-pyran-2-yl)oxy]-chol-5-en-24-oic Acid, TRC

Product Code. 30457255
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457255 10 mg 10mg
30455671 500 mg 500mg
30456555 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457255 Supplier Toronto Research Chemicals Supplier No. TRCT72342510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBDMS-(3beta)-3-[(Tetrahydro-2H-pyran-2-yl)oxy]-chol-5-en-24-oic Acid
Analyte or Component Names TBDMS-(3beta)-3-[(Tetrahydro-2H-pyran-2-yl)oxy]-chol-5-en-24-oic Acid
Shipping Condition Room Temperature
Molecular Formula C35H60O4Si
Synonym (4R)-tert-Butyldimethylsilyl 4-((3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-3-((tetrahydro-2H-pyran-2-yl)oxy)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoate
InChI Formula InChI=1S/C35H60O4Si/c1-24(12-17-31(36)39-40(7,8)33(2,3)4)28-15-16-29-27-14-13-25-23-26(38-32-11-9-10-22-37-32)18-20-34(25,5)30(27)19-21-35(28,29)6/h13,24,26-30,32H,9-12,14-23H2,1-8H3/t24-,26+,27+,28-,29+,30+,32?,34+,35-/m1/s1
Solution Type Neat
SMILES [H][C@@]12CC=C3C[C@H](CC[C@@]3([C@]1(CC[C@]4([C@]2(CC[C@@]4([C@@H](CCC(O[Si](C)(C(C)(C)C)C)=O)C)[H])[H])C)[H])C)OC5CCCCO5
IUPAC Name tert-butyldimethylsilyl (4R)-4-((3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-3-((tetrahydro-2H-pyran-2-yl)oxy)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 572.93
Quantity 10 mg
Formula Weight 572.43
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.