missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBL2 Mouse anti-Human, Clone: 1D5C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66307-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the beta-transducin protein family. Most proteins of the beta-transducin family are involved in regulatory functions. This protein is possibly involved in some intracellular signaling pathway. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.Specifications
| TBL2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y4P3 | |
| TBL2 | |
| TBL2 Fusion Protein Ag24225 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgA |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1D5C2 | |
| Unconjugated | |
| TBL2 | |
| TBL2, transducin (beta) like 2, Transducin beta like protein 2, WBSCR13, WS betaTRP | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26608 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction