Learn More
Invitrogen™ TBPL1 Recombinant Rabbit Monoclonal Antibody (24GB4060)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557282
This item is not returnable.
View return policy
Description
TBPL1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a protein that serves the same function as TBP and substitutes for TBP at some promoters that are not recognized by TFIID. It is essential for spermiogenesis and believed to be important in expression of developmentally regulated genes.
Specifications
| TBPL1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TBPL1 | |
| 21 kDa TBP-like protein; 21-kDA TBP-like protein; 4732475G08; AW011832; AW491032; D18347; MGC:8389; MGC:9620; second TBP of unique DNA protein; STD; STUD; TATA box binding protein-like 1; TATA box binding protein-like protein; TATA box-binding protein related factor 2; TATA box-binding protein-like 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1-like protein; TATA box-binding protein-related factor 2; TATA-box binding protein like 1; tbpl1; TBP-like 1; TBP-like factor; TBP-like protein; TBP-like protein 1; TBP-related factor 2; TBP-related protein; Tlf; TLP; Tlp21; trf; trf2; TRP; Unknown (protein for MGC:134257); zgc:92340 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 689030, 9519 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB4060 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P62380 | |
| TBPL1 | |
| A synthesized peptide derived from human TBPL1 (aa 100-150). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |