Learn More
TBX22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11133-1-AP-150UL
 View more versions of this product
Product Code. 16878003
  • 410.00€ / 150µL
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Mutations in this gene have been associated with the inherited X-linked disorder, Cleft palate with ankyloglossia, and it is believed to play a major role in human palatogenesis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
TBX22
Polyclonal
Unconjugated
TBX22
CLPA, T box 22, T box protein 22, TBOX22, TBX22, TBXX
Rabbit
Antigen Affinity Chromatography
RUO
245572, 302369, 50945
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8K402, Q9Y458
TBX22
TBX22 Fusion Protein Ag1591
150 μL
Primary
Human, Mouse, Rat