missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11133-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Mutations in this gene have been associated with the inherited X-linked disorder, Cleft palate with ankyloglossia, and it is believed to play a major role in human palatogenesis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| TBX22 | |
| Polyclonal | |
| Unconjugated | |
| TBX22 | |
| CLPA, T box 22, T box protein 22, TBOX22, TBX22, TBXX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 245572, 302369, 50945 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8K402, Q9Y458 | |
| TBX22 | |
| TBX22 Fusion Protein Ag1591 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction