missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17825-1-AP-150UL
This item is not returnable.
View return policy
Description
TCEA1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated; transcription, DNA-templated.Specifications
| TCEA1 | |
| Polyclonal | |
| Unconjugated | |
| TCEA1 | |
| GTF2S, SII, SII TFIIS, TCEA, TCEA1, TF2S, TFIIS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21399, 362479, 6917 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P10711, P23193, Q4KLL0 | |
| Tcea1 | |
| TCEA1 Fusion Protein Ag12199 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction