Learn More
Invitrogen™ TCF12 Monoclonal Antibody (690129)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA561740
Product Code. 30535361
  • 429.00€ / 100µg
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
TCF12, also known as HTF4, is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. TCF12 is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. TCF12, in combination with E2A, is required to block thymocyte proliferation prior to pre-TCR expression and is critical for proper T cell differentiation. Recent reports have shown that TCF12 is also a critical factor required for the development of invariant natural killer T cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCF12
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q99081
TCF12
E. coli-derived recombinant human TCF-12/HTF4, Met108-Glu521.
100 μg
Primary
Human
Antibody
IgG1
Immunocytochemistry
690129
Unconjugated
TCF12
A130037E08Rik; ALF1; bHLHb20; Class A helix-loop-helix transcription factor ME1; class B basic helix-loop-helix protein 20; CRS3; DNA-binding protein HTF4; E-box-binding protein; HEB; HEBAlt; helix-loop-helix transcription factor 4; HsT17266; HTF4; HTF-4; ME1; Meb; REB; Salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBPA; SCBP-alpha; TCF12; TCF-12; Transcription factor 12; transcription factor HTF-4
Mouse
Protein A/G
RUO
6938
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized