Learn More
TCF12 Monoclonal Antibody (OTI4D6G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809696S
Description
TCF12 Monoclonal antibody specifically detects TCF12 in Human samples. It is validated for Western BlotSpecifications
| TCF12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99081 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6938 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI4D6G8 | |
| Unconjugated | |
| TCF12 | |
| A130037E08Rik; ALF1; bHLHb20; Class A helix-loop-helix transcription factor ME1; class B basic helix-loop-helix protein 20; CRS3; DNA-binding protein HTF4; E-box-binding protein; HEB; HEBAlt; helix-loop-helix transcription factor 4; HsT17266; HTF4; HTF-4; ME1; Meb; REB; Salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBPA; SCBP-alpha; TCF12; TCF-12; Transcription factor 12; transcription factor HTF-4 | |
| Full length human recombinant protein of human TCF12 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.