missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF4 Antibody (4D4C4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61896
This item is not returnable.
View return policy
Description
TCF4 Monoclonal specifically detects TCF4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| TCF4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| BHLHB19, bHLHb19E2-2, Class B basic helix-loop-helix protein 19, Immunoglobulin transcription factor 2, ITF-2, ITF2PTHS, MGC149724, SEF-2, SEF2-1, SEF2-1A, SEF2-1B, SEF2MGC149723, SL3-3 enhancer factor 2, TCF-4, transcription factor 4 | |
| Mouse | |
| 155 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 4D4C4 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| TCF4 | |
| Purified recombinant fragment of human TCF4 (AA: 518-667) expressed in E. Coli. | |
| Protein G purified | |
| Transcription Factors and Regulators | |
| 6925 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction