missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCL1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47608-25ul
This item is not returnable.
View return policy
Description
TCL1B Polyclonal specifically detects TCL1B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TCL1B | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Oncogene TCL1B, Oncogene TCL-1B, SYN-1, Syncytiotrophoblast-specific protein, T-cell leukemia/lymphoma 1B, T-cell leukemia/lymphoma protein 1B, T-cell lymphoma/leukemia 1B, TCL1, TCL1/MTCP1-like protein 1, TML1TCL1/ MTCP1-like 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TCL1B | |
| This antibody was developed against a recombinant protein corresponding to amino acids: WQMAVHTRELLSSGQMPFSQLPAVWQLYPGRKYRAADSSFWEIADHGQIDSMEQLVLTYQPE | |
| 25 μL | |
| Stem Cell Markers | |
| 9623 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction