missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR alpha/beta Antibody (BMA031) [PE], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43550PE
This item is not returnable.
View return policy
Description
TCR alpha/beta Monoclonal antibody specifically detects TCR alpha/beta in Human samples. It is validated for Immunoprecipitation,Neutralization,Serum,Immunocytochemistry/ Immunofluorescence
Specifications
| TCR alpha/beta | |
| Monoclonal | |
| PE | |
| C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
| The original antibody was generated by immunizing mice with human perihperal blood T-lymphocytes. | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 6955 | |
| Store at 4⪚C in the dark. | |
| IgG |
| Immunoprecipitation, Neutralization, Immunocytochemistry/Immunofluorescence | |
| BMA031 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction