missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR beta Armenian Hamster, FITC, Clone: H57-597, enQuireBio™
Armenian Hamster Monoclonal Antibody
85.85€ - 250.00€
Specifications
Antigen | TCR beta |
---|---|
Clone | H57-597 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15912498
|
EnquireBio
QAB79-F-25UG |
25 μg |
85.85€
25µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15902498
|
EnquireBio
QAB79-F-100UG |
100 μg |
98.45€
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922498
|
EnquireBio
QAB79-F-500UG |
500 μg |
250.00€
500µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TCR beta | |
0.5mg/mL | |
Monoclonal | |
Armenian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Tcrb | |
Primary | |
Store at 2-4°C. |
H57-597 | |
Flow Cytometry | |
FITC | |
RUO | |
21577 | |
IgG | |
Protein A/G Purified |