missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR beta Armenian Hamster, PerCP-Cy5.5, Clone: H57-597, enQuireBio™
Armenian Hamster Monoclonal Antibody
169.05€ - 367.50€
Specifications
| Antigen | TCR beta |
|---|---|
| Clone | H57-597 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15942498
|
EnquireBio
QAB79-PCP55-25ug |
25 μg |
169.05€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15932498
|
EnquireBio
QAB79-PCP55-100ug |
100 μg |
367.50€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| TCR beta | |
| 0.5mg/mL | |
| Monoclonal | |
| Armenian Hamster | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| Tcrb | |
| Primary | |
| Store at 2-4°C. |
| H57-597 | |
| Flow Cytometry | |
| PerCP-Cyanine5.5 | |
| RUO | |
| 21577 | |
| IgG | |
| Protein A/G Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title