missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR Vα24-Jα18 (iNKT cell) Mouse anti-Human, BUV563, Clone: 6B11, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 748830
This item is not returnable.
View return policy
Description
TCR Vα24-Jα18 (iNKT cell) Monoclonal antibody specifically detects TCR Vα24-Jα18 (iNKT cell) in Human samples. It is validated for Flow Cytometry
Specifications
| TCR Vα24-Jα18 (iNKT cell) | |
| Monoclonal | |
| Brilliant Ultraviolet 563 | |
| Vα24JαQ TCR Chain | |
| Peptide from CDR3 loop of the invariant TCR V24 sequence | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 6B11 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction