missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR V beta 8a Antibody (I6G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22502-50ug
This item is not returnable.
View return policy
Description
TCR V beta 8a Monoclonal specifically detects TCR V beta 8a in Human, Mouse, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Immunofluorescence.
Specifications
| TCR V beta 8a | |
| Monoclonal | |
| 0.15 mg/mL | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunofluorescence | |
| T cell receptor beta locus, T-cell antigen receptor, beta polypeptide, T-cell receptor, beta cluster, T-cell receptor, beta cluster, TCRB, TRB | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6957 | |
| Human, Mouse, Primate | |
| Ascites |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunofluorescence | |
| I6G8 | |
| Unconjugated | |
| P04435 | |
| TCB | |
| Human TCR V beta 8 | |
| 50 μg | |
| Primary | |
| This antibody reacts with Human and Primate. | |
| Store at 4°C. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction