missing translation for 'onlineSavingsMsg'
Learn More

TCS 5861528, Tocris Bioscience™

Product Code. 18075932
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18075932 50 mg 50mg
18024782 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18075932 Supplier Tocris Bioscience Supplier No. 3938/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TRPA1 blocker

TRPA1 channel blocker that antagonizes AITC- and 4-HNE-evoked calcium influx (IC50 values are 14.3 and 18.7μM respectively). Attenuates diabetic hypersensitivity in an in vivo rat model.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332117-28-9
Molecular Formula C19H23N5O3
Molecular Weight (g/mol) 369.425
InChI Key ZUTUWJYMCADJHD-UHFFFAOYSA-N
Synonym chembridge-5861528, n-4-butan-2-ylphenyl-2-1,3-dimethyl-2,6-dioxopurin-7-yl acetamide, 2-1,3-dimethyl-2,6-dioxopurin-7-yl-n-4-sec-butyl phenyl acetamide, d0j0bv, n-4-2r-butan-2-yl phenyl-2-1,3-dimethyl-2,6-dioxopurin-7-yl acetamide, n-4-sec-butylphenyl-2-1,3-dimethyl-2,6-dioxo-2,3-dihydro-1h-purin-7 6h-yl acetamide, 2-1,3-dimethyl-2,6-dioxo 1,3,7-trihydropurin-7-yl-n-4-methylpropyl phenyl acetamide, 2-1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7h-purin-7-yl-n-4-1-methylpropyl phenyl acetamide, n-4-butan-2-yl phenyl-2-1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7h-purin-7-yl acetamide, n-4-butan-2-yl phenyl-2-1,3-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1h-purin-7-yl acetamide
PubChem CID 2873523
IUPAC Name N-(4-butan-2-ylphenyl)-2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetamide
SMILES CCC(C)C1=CC=C(C=C1)NC(=O)CN2C=NC3=C2C(=O)N(C(=O)N3C)C

Specifications

Quantity 50 mg
Formula Weight 369.42
Percent Purity >98%
Chemical Name or Material TCS 5861528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.