missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTN3 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97323H
This item is not returnable.
View return policy
Description
TCTN3 Polyclonal antibody specifically detects TCTN3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TCTN3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| HRP | |
| C10orf61, chromosome 10 open reading frame 61, DKFZP564D116, TECT3DKFZp564D116, tectonic 3, tectonic family member 3, tectonic-3 | |
| Produced in rabbits immunized with E. coli-derived Human TCTN3 fragment. | |
| 0.1 mL | |
| Apoptosis, Cell Biology | |
| 26123 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction