missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20627
This item is not returnable.
View return policy
Description
TDG Polyclonal specifically detects TDG in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| TDG | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.01% Thimerosal | |
| EC 3.2.2.29, thymine-DNA glycosylaseG/T mismatch-specific thymine DNA glycosylase | |
| Rabbit | |
| 46 kDa | |
| 0.1 mL | |
| Base Excision Repair, DNA Repair | |
| 6996 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| 1mg/mL | |
| Western Blot 1:5000-1:20000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Knockout Validated | |
| Q13569 | |
| TDG | |
| Recombinant protein encompassing a sequence within the center region of human TDG. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction