missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDO2 Antibody (998604) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9768JF525
This item is not returnable.
View return policy
Description
TDO2 Monoclonal antibody specifically detects TDO2 in Human samples. It is validated for Immunohistochemistry,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| TDO2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| TDOTO, TPH2EC 1.13.11.11, TRPOtryptophanase, Tryptamin 2,3-dioxygenase, tryptophan 2,3-dioxygenase, Tryptophan oxygenase, Tryptophan pyrrolase, Tryptophanase | |
| E. coli-derived human TDO2, Leu18-Phe388, Accession # P48775 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 998604 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology, Lipid and Metabolism, Neuroscience | |
| 6999 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction