missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDRD3 Antibody (RAB-C419), Alexa Fluor™ 700 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20149AF700
This item is not returnable.
View return policy
Description
TDRD3 Monoclonal antibody specifically detects TDRD3 in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
| TDRD3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| FLJ21007, tudor domain containing 3, tudor domain-containing protein 3 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing TDRD3 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Neurodegeneration, Neuroscience | |
| 81550 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunofluorescence | |
| RAB-C419 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction