missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TDRKH Monoclonal Antibody (686213)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561646
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
TDRKH is a protein of unknown function containing one tudor domain, and two KH domains. It has been found to interact with PIWIL1 and PIWIL4 by binding to methylated arginines residues.
Specifications
| TDRKH | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9Y2W6 | |
| TDRKH | |
| E. coli-derived recombinant human TDRKH, His206-Leu561. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Frozen) | |
| 686213 | |
| Unconjugated | |
| TDRKH | |
| 2700091C21Rik; putative RNA binding protein; RGD1311196; RP11-98D18.8; Tdrd2; TDRD7; Tdrkh; tudor and KH domain containing; tudor and KH domain containing protein; tudor and KH domain-containing protein; tudor domain containing 2; tudor domain-containing protein 2 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 11022 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction