Learn More
TDRKH Mouse anti-Human, Unconjugated, Clone: 2B1B12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66845-1-IG-20UL
Description
The TDRKH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TDRKH. This antibody recognizes human, mouse, rat, pig antigen. The TDRKH antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| TDRKH | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunofluorescence, ELISA | |
| 2B1B12 | |
| Unconjugated | |
| 20 μL | |
| 11022 | |
| IgG1 |