Learn More
TDRKH Mouse anti-Human, Unconjugated, Clone: 2B1B12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66845-1-IG-20UL
 View more versions of this product
Product Code. 16829095
  • 145.00€ / 20µL
Description
The TDRKH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TDRKH. This antibody recognizes human, mouse, rat, pig antigen. The TDRKH antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
TDRKH
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunofluorescence, ELISA
2B1B12
Unconjugated
20 μL
11022