missing translation for 'onlineSavingsMsg'
Learn More

Tebuthiuron, TRC

Product Code. 30413362
Change view
Click to view available options
Quantity:
250 mg
2.5 g
25 g
Unit Size:
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30413362 2.5 g 2.50g
30412814 250 mg -
30409810 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30413362 Supplier Toronto Research Chemicals Supplier No. TRCT0136202.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34014-18-1
Molecular Formula C9 H16 N4 O S
Molecular Weight (g/mol) 228.31
Synonym Spike, N-[5-(1,1-dimethylethyl)-1,3,4-thiadiazol-2-yl]-N,N′-dimethylurea
IUPAC Name 1-(5-tert-butyl-1,3,4-thiadiazol-2-yl)-1,3-dimethylurea
SMILES CNC(=O)N(C)c1nnc(s1)C(C)(C)C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H16N4OS/c1-9(2,3)6-11-12-8(15-6)13(5)7(14)10-4/h1-5H3,(H,10,14)
Formula Weight 228.1045
Chemical Name or Material Tebuthiuron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.