Learn More
Unity Lab Services™ Tech Direct Service Plan for Thermo Scientific Bio Safety Cabinets
Description
- Continue service support post factory warranty with enhanced remote support and annual preventative maintenance
- 3 business day response time (may vary by location) for corrective maintenance
- 15% discount off billable rates for engineer labor, and factory-certified replacement parts for on-site corrective maintenance visit
- One scheduled Preventative Maintenance visit, including a preventative maintenance kit (where applicable)
- Priority unlimited access to OEM-trained Technical support experts who can diagnose, investigate, and remotely resolve issues or schedule on-site visits using the latest digital remote support tools
- Access to on-line service knowledge base
- Corrective Maintenance Repair Guarantee for the duration of product warranty or 1 year whichever is longer, as defined in the contract terms and conditions
- Instrument Software/ Firmware updates included as needed to support a preventative maintenance visit
- Upgrades to new versions of software are not included
- 10% discount on additional services including additional preventative maintenance visits, calibration, and qualification services
Specifications
Specifications
| Duration | 1 Year |
| For Use With (Equipment) | Biosafety Cabinet (1.8 m) |
| For Use With (Application) | Support Post-Factory Warranty |
| Warranty and Service Offering | Priority Technical Support, One Preventative Maintenance Visit |
| Includes | 3 Day Response Time (Within Specified Location and Criteria), Priority Technical Support, and One Preventative Maintenance Visit |
| Product Type | Tech Direct Service Plan |
| Services Not Included | Upgrades to New Software Versions |
| Warranty Response Time | 3 Days (Within Specified Location and Criteria) |
| Services Included | Priority Technical Support, and One Preventative Maintenance Visit |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?