missing translation for 'onlineSavingsMsg'
Learn More

Tedalinab, TRC

Product Code. 30421436
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421436 10 mg 10mg
30411881 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30421436 Supplier Toronto Research Chemicals Supplier No. TRCT01373010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916591-01-0
Molecular Formula C19H21F2N3O
Molecular Weight (g/mol) 345.39
Synonym (4S,7R)-1-(2,4-Difluorophenyl)-N-(1,1-dimethylethyl)-4,5,6,7-tetrahydro-4,7-methano-1H-indazole-3-carboxamide
SMILES CC(C)(C)NC(=O)c1nn(c2[C@@H]3CC[C@@H](C3)c12)c4ccc(F)cc4F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H21F2N3O/c1-19(2,3)22-18(25)16-15-10-4-5-11(8-10)17(15)24(23-16)14-7-6-12(20)9-13(14)21/h6-7,9-11H,4-5,8H2,1-3H3,(H,22,25)/t10-,11+/m0/s1
Formula Weight 345.1653
Chemical Name or Material Tedalinab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.