missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEF1 Rabbit anti-Human, Mouse, Rat, Clone: S05-1H3, Novus Biologicals™
Rabbit Monoclonal Antibody
286.00€ - 581.00€
Specifications
| Antigen | TEF1 |
|---|---|
| Clone | S05-1H3 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18391682
|
Bio-Techne
NBP3-15100-25UL |
25 μg |
286.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18375573
|
Bio-Techne
NBP3-15100-100UL |
100 μg |
581.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TEF1 Monoclonal antibody specifically detects TEF1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
| TEF1 | |
| Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| NTEF-1, protein GT-IIC, TEA domain family member 1 (SV40 transcriptional enhancer factor), TEAD-1 | |
| A synthetic peptide of human TEF1 (Uniprot#: P28347) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S05-1H3 | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Cell Biology | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 7003 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title