Learn More
TEFM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554887
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RRSINIVEHR ENFGPFQNLE SLMNVPLFKY KSTVQVCNSI LCPKTGREKR KSPENRFLRK LLKPDIERER LKAVNSIISI V Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.
TEFM is a transcription elongation factor which increases mitochondrial RNA polymerase processivity. It regulates transcription of the mitochondrial genome, including genes important for the oxidative phosphorylation machinery.Specifications
TEFM | |
Polyclonal | |
Unconjugated | |
TEFM | |
1110002N22Rik; C17orf42; C9H17orf42; RGD1306595; Tefm; transcription elongation factor of mitochondria; transcription elongation factor, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79736 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96QE5 | |
TEFM | |
Recombinant protein corresponding to Human TEFM | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only