missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMILIN1 Antibody (C11A8), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-36338PCP
This item is not returnable.
View return policy
Description
EMILIN1 Monoclonal antibody specifically detects EMILIN1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| EMILIN1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| 11117 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
| C11A8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen | |
| DKFZp586M121, elastin microfibril interface-located protein 1, elastin microfibril interfacer 1, EMI, EMILIN, EMILIN-1, gp115 | |
| recombinant mouse Emilin-1 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction