Learn More
Thermo Scientific™ Temperature Chart Recorder Paper
Product Code. 11888773 Shop All Thermo Scientific Products
Change view
Click to view available options
Temperature Range:
-200°C to 0°C
-40°C to +25°C
Unit Size:
Each
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range Unit Size
11888773 -40°C to +25°C Pack of 50
10354557 -200°C to 0°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11888773 Supplier Thermo Scientific™ Supplier No. 6184
  • ONLINE EXCLUSIVE PRICE
    81.85€
    76.40€ / Pack of 50
    Save 5.45€
    6% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.
Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.
  • 6' (15.2cm) diameter
  • For use in freestanding and panel-mounted chart recorders
TRUSTED_SUSTAINABILITY
Specifications
Type Chart Recorder Paper
Temperature Range -40°C to +25°C
For Use With (Equipment) Free-Standing and Panel-Mounted Chart Recorders
Monitoring Options Recorder
Static Holding Time 7 Days
Diameter (English) 6 in.
Diameter (Metric) 15.2 cm