missing translation for 'onlineSavingsMsg'
Learn More

Horst™ Temperature Controllers

Product Code. 10047131
Change view
Click to view available options
Model:
HT 30
HT 31
HT30
HT31
Sensor Types:
NiCr-Ni (K)
Pt 100
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Sensor Types unitSize
10047131 HT30 NiCr-Ni (K) Each
11908325 HT31 NiCr-Ni (K) Each
10352222 HT 30 Pt 100 Each
10502702 HT 31 Pt 100 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10047131 Supplier Horst™ Supplier No. 063001

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Wattage 2200 or 3500w
Power Density 230 V - 50 Hz
Model HT30
Sensor Types NiCr-Ni (K)
Amperage 10 A
Type Temperature Controller
Voltage 230 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.