Learn More
Horst™ Temperature Controllers
Specifications
Specifications
| Wattage | 3500w |
| Power Density | 3600 |
| Model | HT31 |
| Sensor Types | NiCr-Ni (K) |
| Amperage | 16 A |
| Type | Temperature Controller |
| Voltage | 230 V |