Learn More
Dostmann Electronic™ Immersion Temperature Probe
Product Code. 10661366
Change view
Click to view available options
Length (Metric):
120 mm
130 mm
220 mm
300 mm
Temperature Range (Metric):
-100°C to +1100°C
-200°C to +1100°C
-40°C to +400°C
Diameter (Metric) Probe:
1 mm
1.5 mm
3 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Temperature Range (Metric) Diameter (Metric) Probe unitSize
10661366 300 mm -100°C to +1100°C 3 mm Each
10559625 300 mm -100°C to +1100°C 1.5 mm Each
10335548 220 mm -200°C to +1100°C 3 mm Each
11894392 120 mm -40°C to +400°C 3 mm Each
11836850 130 mm -200°C to +1100°C 1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
R\u00E9cupel-bijdrage uitgesloten
Product Code. 10661366 Supplier Dostmann Electronic™ Supplier No. 60100503
Only null left
Add to basket
Add to basket
R\u00E9cupel-bijdrage uitgesloten
Dostmann Electronic™ Immersion Temperature Probe is useful for temperature measurements in liquids and powdered materials.
  • Type K according to EN 60584-1
  • NiCr-Ni
  • Mineral-insulated
Specifications
Diameter (Metric) Probe 3 mm
For Use With (Application) For Measuring In Liquid And Powdered Materials
Includes Handle and cable
Length (Metric) 300 mm
Product Type Temperature Probe
Description Non-flexible probe
Temperature Range (Metric) -100°C to +1100°C
For Use With (Equipment) P410 / TC301 / TC 309 / Mini-K