missing translation for 'onlineSavingsMsg'
Learn More

Mapa Professional TempTec 332 Thermal Protection Glove

Product Code. 30533544
Change view
Click to view available options
Size:
8
9
10
Unit Size:
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30533544 9 1 pair
30533546 8 1 pair
30533551 10 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30533544 Supplier Mapa Professional Supplier No. 34332029
Only null left
Add to basket
Edge
Add to basket

Effective thermal insulation and resistance to a wide range of chemicals.

ISO cut level: X

Productivity: Good grip on slippery objects, due to the embossed texture.

Guaranteed quality: Silicone free to prevent traces, defects on metal sheet and glass prior to painting.

Thermal protection: Excellent thermal insulation, due to double-knit cotton.

Skin protection: Multi-purpose chemical resistance (acids, aliphatic solvents) increased by the thickness of the material.

Specifications

ANSI Abrasion Rating ANSI 0
Certifications/Compliance (Europe) CE Category 3
Chlorinated No
Applications Industrial
Color Dark Blue
Latex Free Yes
Material Polychloroprene
Product Type Neoprene Gloves
Size 9
EN388 Standard EN388:2212X
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES KPT
Glove Cuff Style Open
Grip Design Embossed
Liner Material Knitted thermal protection
Lining Thermal Lining
Packaging Type Individual Bags
Resistant To Chemicals
Length (Metric) 36 cm
Product Line TempTec 332
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.