missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenascin C Rabbit anti-Human, Clone: TNC/2981R, Novus Biologicals™
Rabbit Monoclonal Antibody
309.00€
Specifications
| Antigen | Tenascin C |
|---|---|
| Clone | TNC/2981R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 2-4 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Tenascin C Monoclonal specifically detects Tenascin C in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Tenascin C | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 3371 | |
| IgG | |
| Protein A or G purified |
| TNC/2981R | |
| Immunohistochemistry-Paraffin 2-4 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP | |
| Recombinant human full-length Tenascin C protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title