Learn More
Tensin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557515
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LSPLTNGLDK SYPMEPMVNG GGYPYESASR AGPAHAGHTA PMRPSYSAQE GLAGYQREGP HPAWPQPVTT SHYAHDPSGM FRSQS Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
Tensin is involved in the maintenance of cellular structure by anchoring Actin filaments at the focal adhesion via F-Actin binding and capping activities. However, tensin also contains a Src homology 2 (SH2) domain and has the ability to be phosphorylated. Tensin is phosphorylated on tyrosine, serine, and threonine residues, suggesting that it might participate in signal transduction cascades. These diverse characteristics in a single molecule indicate that tensin may be an important link between the cytoskeleton and signal transduction pathways.Specifications
Tensin 1 | |
Polyclonal | |
Unconjugated | |
TNS1 | |
1110018I21Rik; 1200014E20Rik; AI648117; DKFZp586K0617; E030018G17Rik; E030037J05Rik; matrix-remodelling associated 6; Matrix-remodelling-associated protein 6; MGC88584; MST091; MST122; MST127; MSTP091; MSTP122; MSTP127; MXRA6; PPP1R155; protein phosphatase 1, regulatory subunit 155; tensin 1; tensin-1; TNS; TNS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7145 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBL0 | |
TNS1 | |
Recombinant protein corresponding to Human Tensin 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only