missing translation for 'onlineSavingsMsg'
Learn More

TER-119 Rat, PE, Clone: TER-119, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB76-PE-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15922448

  • 263.00€ / 100µg
Estimated Shipment: 08-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TER-119
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Rat
100 μg
Primary
Store at 2-4°C.
Flow Cytometry
TER-119
PE
Ly76
Protein A/G Purified
RUO
104231
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions