missing translation for 'onlineSavingsMsg'
Learn More

Terbutaline, TRC

Product Code. 17912785
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17912785 50 mg 50mg
17924035 100 mg 100mg
17903395 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17912785 Supplier Toronto Research Chemicals Supplier No. TRCT10976350MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23031-25-6
Molecular Formula C12 H19 N O3
Molecular Weight (g/mol) 225.28
Synonym 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, Benzyl alcohol, α-[(tert-butylamino)methyl]-3,5-dihydroxy- (8CI), 5-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-1,3-benzenediol, (±)-Terbutaline, 1-(3,5-Dihydroxyphenyl)-2-(tert-butylamino)ethanol, AR-D 080301, AR-DO80301, Aerodur, Brican, Bricar, Bricaril, Bricyn, Fasma, KWD 2019, Terbutalin, Terbutaline
IUPAC Name 5-[2-(tert-butylamino)-1-hydroxyethyl]benzene-1,3-diol
SMILES CC(C)(C)NCC(O)c1cc(O)cc(O)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H19NO3/c1-12(2,3)13-7-11(16)8-4-9(14)6-10(15)5-8/h4-6,11,13-16H,7H2,1-3H3
Formula Weight 225.1365
Percent Purity >95%
Chemical Name or Material Terbutaline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.