missing translation for 'onlineSavingsMsg'
Learn More

Terbuthylazine, TRC

Product Code. 17914355
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914355 250 mg 250mg
17950895 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17914355 Supplier Toronto Research Chemicals Supplier No. TRCT110500250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5915-41-3
Molecular Formula C9 H16 Cl N5
Molecular Weight (g/mol) 229.71
Synonym Terbuthylazine, 1,3,5-Triazine-2,4-diamine, 6-chloro-N2-(1,1-dimethylethyl)-N4-ethyl- (ACI), 1,3,5-Triazine-2,4-diamine, 6-chloro-N-(1,1-dimethylethyl)-N'-ethyl- (9CI), 6-Chloro-N2-(1,1-dimethylethyl)-N4-ethyl-1,3,5-triazine-2,4-diamine (ACI), s-Triazine, 2-(tert-butylamino)-4-chloro-6-(ethylamino)- (7CI, 8CI), 2-Chloro-4-(ethylamino)-6-(tert-butylamino)-s-triazine, 2-Chloro-4-ethylamino-6-tert-butylamino-1,3,5-triazine, 2-Chloro-4-tert-butylamino-6-ethylamino-s-triazine, 2-N-tert-Butyl-6-chloro-4-N-ethyl-1,3,5-triazine-2,4-diamine, 2-tert-Butylamino-4-chloro-6-ethylamino-s-triazine, 4-Ethylamino-6-tert-butylamino-2-chloro-S-triazine, 6-Chloro-2-tert-butylamino-4-(ethylamino)-[1,3,5]-triazine, ChlorCaragard, Click, Click 500SC, G 13529, Gardeprim A 1862, Gardoprim, GS 13529, N-tert-Butyl-6-chloro-N'-ethyl-1,3,5-triazine-2,4-diamine, Primatol M, Terbutazine, Terbutylazine, Terbutylethylazine, tert-Butylazine, Tyllanex
IUPAC Name 2-N-tert-butyl-6-chloro-4-N-ethyl-1,3,5-triazine-2,4-diamine
SMILES CCNc1nc(Cl)nc(NC(C)(C)C)n1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H16ClN5/c1-5-11-7-12-6(10)13-8(14-7)15-9(2,3)4/h5H2,1-4H3,(H2,11,12,13,14,15)
Formula Weight 229.1094
Chemical Name or Material Terbuthylazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.