missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene-8-carboxylate, Thermo Scientific™

Product Code. 11916671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11916671 1 g Glass bottle 1g
11926671 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916671 Supplier Thermo Scientific Acros Supplier No. 438590010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 185099-68-7
Molecular Formula C13H18F3NO5S
Molecular Weight (g/mol) 357.35
InChI Key CJDKVEWDWXIBSM-UHFFFAOYSA-N
Synonym 8-boc-3-trifluoromethylsulfonyloxy-8-azabicyclo 3.2.1 oct-3-ene, tert-butyl 3-trifluoromethanesulfonyloxy-8-azabicyclo 3.2.1 oct-2-ene-8-carboxylate, tert-butyl 3-trifluoromethylsulfonyloxy-8-azabicyclo 3.2.1 oct-2-ene-8-carboxylate, tert-butyl 3-trifluoromethylsulfonyloxy-8-azabicyclo 3.2.1 oct-3-ene-8-carboxylate, 8-azabicyclo 3.2.1 oct-2-ene-8-carboxylic acid,3-trifluoromethyl sulfonyl oxy-, 1,1-dimethylethyl ester, tert-butyl 3-trifluoromethyl sulfonyl oxy-8-azabicyclo 3.2.1 oct-2-ene-8-carboxylate, 1r,5s-tert-butyl 3-trifluoromethylsulfonyloxy, tert-butyl 3-trifluoromethyl sulfonyl oxy-8-azabicyclo 3.2.1 oct-3-ene-8-carboxylate, tert-butyl 3-trifluoromethanesulfonyl oxy-8-azabicyclo 3.2.1 oct-2-ene-8-carboxylate
PubChem CID 22648094
IUPAC Name tert-butyl 3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene-8-carboxylate
SMILES CC(C)(C)OC(=O)N1C2CCC1C=C(C2)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 36°C to 40°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 357.35
Percent Purity ≥94.0%
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material tert-Butyl 3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene-8-carboxylate
compliance-icons
Product Identifier
  • tert-Butyl 3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene-8-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.