missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-((tosyloxy)methyl)piperidine-1-carboxylate, TRC

Product Code. 30334483
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30334483

Brand: Toronto Research Chemicals TRCB80955310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl 4-((tosyloxy)methyl)piperidine-1-carboxylate
CAS 166815-96-9
Molecular Formula C18 H27 N O5 S
Quantity 10 mg
Synonym 1-(tert-Butoxycarbonyl)-4-(4-methylphenylsulfonyloxymethyl)piperidine, 1-(tert-Butoxycarbonyl)-4-[[(p-tolylsulfonyl)oxy]methyl]piperidine, 4-(((Toluene-4-sulfonyl)oxy)methyl)piperidine-1-carboxylic Acid tert-Butyl Ester, 4-(4-Methylphenylsulfonyloxymethyl)-1-tert-butoxycarbonylpiperidine, 4-[[(4-Tolylsulfonyl)oxy]methyl]piperidine-1-carboxylic Acid tert-Butyl Ester, 4-[[[(4-Methylphenyl)sulfonyl]oxy]methyl]-1-piperidinecarboxylic Acid 1,1-Dimethylethyl Ester, N-(tert-Butoxycarbonyl)-4-(tosyloxymethyl)piperidine, N-tert-Butoxycarbonyl-4-(4-toluenesulfonyloxymethyl)piperidinetert-Butyl 4-(p-tolylsulfonyloxymethyl)piperidine-1-carboxylate, tert-Butyl 4-(tosyloxymethyl)piperidine-1-carboxylate, tert-Butyl 4-[[[(4-Methylphenyl)sulfonyl]oxy]methyl]piperidine-1-carboxylate
InChI Formula InChI=1S/C18H27NO5S/c1-14-5-7-16(8-6-14)25(21,22)23-13-15-9-11-19(12-10-15)17(20)24-18(2,3)4/h5-8,15H,9-13H2,1-4H3
SMILES Cc1ccc(cc1)S(=O)(=O)OCC2CCN(CC2)C(=O)OC(C)(C)C
IUPAC Name tert-butyl 4-[(4-methylphenyl)sulfonyloxymethyl]piperidine-1-carboxylate
Molecular Weight (g/mol) 369.476
Formula Weight 369.161
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.