missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-((tosyloxy)methyl)piperidine-1-carboxylate, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | tert-Butyl 4-((tosyloxy)methyl)piperidine-1-carboxylate |
| CAS | 166815-96-9 |
| Molecular Formula | C18 H27 N O5 S |
| Quantity | 50 mg |
| Synonym | 1-(tert-Butoxycarbonyl)-4-(4-methylphenylsulfonyloxymethyl)piperidine, 1-(tert-Butoxycarbonyl)-4-[[(p-tolylsulfonyl)oxy]methyl]piperidine, 4-(((Toluene-4-sulfonyl)oxy)methyl)piperidine-1-carboxylic Acid tert-Butyl Ester, 4-(4-Methylphenylsulfonyloxymethyl)-1-tert-butoxycarbonylpiperidine, 4-[[(4-Tolylsulfonyl)oxy]methyl]piperidine-1-carboxylic Acid tert-Butyl Ester, 4-[[[(4-Methylphenyl)sulfonyl]oxy]methyl]-1-piperidinecarboxylic Acid 1,1-Dimethylethyl Ester, N-(tert-Butoxycarbonyl)-4-(tosyloxymethyl)piperidine, N-tert-Butoxycarbonyl-4-(4-toluenesulfonyloxymethyl)piperidinetert-Butyl 4-(p-tolylsulfonyloxymethyl)piperidine-1-carboxylate, tert-Butyl 4-(tosyloxymethyl)piperidine-1-carboxylate, tert-Butyl 4-[[[(4-Methylphenyl)sulfonyl]oxy]methyl]piperidine-1-carboxylate |
| InChI Formula | InChI=1S/C18H27NO5S/c1-14-5-7-16(8-6-14)25(21,22)23-13-15-9-11-19(12-10-15)17(20)24-18(2,3)4/h5-8,15H,9-13H2,1-4H3 |
| SMILES | Cc1ccc(cc1)S(=O)(=O)OCC2CCN(CC2)C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl 4-[(4-methylphenyl)sulfonyloxymethyl]piperidine-1-carboxylate |
| Molecular Weight (g/mol) | 369.476 |
| Formula Weight | 369.161 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction