missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Bromoacetate, TRC

Product Code. 30338987
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338987 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30338987 Supplier Toronto Research Chemicals Supplier No. TRCB69074510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyl Bromoacetate
CAS 5292-43-3
Molecular Formula C6H11BrO2
Quantity 10 g
Synonym 1,1-Dimethylethyl 2-Bromoacetate, 1,1-Dimethylethyl bromoacetate, 1,1-Dimethylethyl monobromoacetate, 2-Bromoacetic Acid tert-Butyl Ester, Bromoacetic Acid 1,1-Dimethylethyl Ester, Bromoacetic Acid tert-Butyl Ester, NSC 82470, tert-Butyl 2-Bromoacetate, tert-Butyl Bromoacetate, tert-Butyl alpha-Bromoacetate, t-Butylbromoacetate
InChI Formula InChI=1S/C6H11BrO2/c1-6(2,3)9-5(8)4-7/h4H2,1-3H3
SMILES CC(C)(C)OC(=O)CBr
IUPAC Name tert-butyl 2-bromoacetate
Molecular Weight (g/mol) 195.05
Formula Weight 193.9942
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.