Learn More
tert-Butyl hydrogen phthalate, 97%
Description
It finds its application in the resolution and reactions of tertiary alcohols : 2-phenylbutan-2-ol and 3-methylhexan-3-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It finds its application in the resolution and reactions of tertiary alcohols : 2-phenylbutan-2-ol and 3-methylhexan-3-ol.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 84°C to 88°C |
| Quantity | 25 g |
| Beilstein | 2263386 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 222.24 |
| Percent Purity | 97% |
| Chemical Name or Material | tert-Butyl hydrogen phthalate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.