Learn More
tert-Butyl N-((1S)-1-Benzyl-3-chloro-2-oxopropyl)carbamate, Mikromol™
Product Code. 30052274
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052274 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052274 Supplier Mikromol Supplier No. MM3624.010025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
tert-Butyl N-((1S)-1-Benzyl-3-chloro-2-oxopropyl)carbamate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material tert-Butyl N-((1S)-1-Benzyl-3-chloro-2-oxopropyl)carbamate
CAS 102123-74-0
Analyte or Component Names tert-Butyl N-((1S)-1-Benzyl-3-chloro-2-oxopropyl)carbamate
Shipping Condition Room Temperature
Molecular Formula C15 H20 Cl N O3
Certifications/Compliance ISO 9001
Synonym Carbamic acid, N-[(1S)-3-chloro-2-oxo-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester, Carbamic acid, [(1S)-3-chloro-2-oxo-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester (9CI), Carbamic acid, [3-chloro-2-oxo-1-(phenylmethyl)propyl]-, 1,1-dimethylethyl ester, (S)-, (3S)-(-)-3-tert-Butoxycarbonylamino-1-chloro-4-phenyl-2-butanone, (3S)-3-(tert-Butoxycarbonylamino)-1-chloro-4-phenyl-2-butanone, N-Boc phenylalanyl chloride
InChI Formula InChI=1S/C15H20ClNO3/c1-15(2,3)20-14(19)17-12(13(18)10-16)9-11-7-5-4-6-8-11/h4-8,12H,9-10H2,1-3H3,(H,17,19)/t12-/m0/s1
SMILES CC(C)(C)OC(=O)N[C@@H](Cc1ccccc1)C(=O)CCl
IUPAC Name tert-butyl N-[(2S)-4-chloro-3-oxo-1-phenylbutan-2-yl]carbamate
Molecular Weight (g/mol) 297.7772
Quantity 25 mg
Formula Weight 297.1132 g/mol