Learn More
tert-Butyl propiolate, 98%
Product Code. 10341180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10341180 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10341180 Supplier Thermo Scientific Acros Supplier No. 435390010
 Restricted Item
Estimated Shipment: 08-07-2027
Add to basket
missing translation for 'validMainframeMsg'
CAS: 13831-03-3 | C7H10O2 | 126.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 13831-03-3
Melting Point 18°C to 20°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 52°C to 53°C (27.0 mmHg)
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H10O2
MDL Number MFCD00060100
Quantity 1 g
Specific Gravity 0.92
Synonym tert-butyl propiolate, propiolic acid tert-butyl ester, t-butyl propiolate, 2-propynoic acid, 1,1-dimethylethyl ester, boc-acetylene, tert-butyl propalate, tert-butyl propargylate, acmc-209chj, tert-butyl propiolate #
InChI Key XGTPDIIFEPTULX-UHFFFAOYSA-N
SMILES CC(C)(C)OC(=O)C#C
IUPAC Name tert-butyl prop-2-ynoate
Molecular Weight (g/mol) 126.16
PubChem CID 543038
Formula Weight 126.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butyl propiolate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl propiolate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.