missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylsilyl Chloride, TRC

Product Code. 30339237
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30339237 Supplier Toronto Research Chemicals Supplier No. TRCB6910805G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material tert-Butyldimethylsilyl Chloride
CAS 18162-48-6
Purity Grade Notes GC
Molecular Formula C6 H15 Cl Si
Quantity 5 g
Synonym (1,1-Dimethylethyl)dimethylsilyl chloride, 2-(Chlorodimethylsilyl)-2-methylpropane, Chloro(1,1-dimethylethyl)dimethylsilane, Chloro-tert-butyldimethylsilane, Chlorodimethyl-tert-butylsilane, Dimethyl(1,1-dimethylethyl)chlorosilane, Dimethyl-tert-butylchlorosilane, Dimethyl-tert-butylsilyl chloride, LS 1190, TBDMS chloride, TBDMS-Cl, TBDMSCl, TBSCl, t-Butyldimethylchlorosilane, tert-Butyl(dimethyl)silane chloride, tert-Butylchlorodimethylsilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, Chloro(1,1-dimethylethyl)dimethyl-silane, TBDMS, TBS-Cl
InChI Formula InChI=1S/C6H15ClSi/c1-6(2,3)8(4,5)7/h1-5H3
SMILES CC(C)(C)[Si](C)(C)Cl
IUPAC Name tert-butyl-chloro-dimethylsilane
Molecular Weight (g/mol) 150.72
Formula Weight 150.0632
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.