missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldiphenylchlorosilane, TRC

Product Code. 30178970
Change view
Click to view available options
Quantity:
2 g
10 g
25 g
Unit Size:
10g
25g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30178970 10 g 10g
30179517 2 g 2g
30179031 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30178970 Supplier Toronto Research Chemicals Supplier No. TRCB69241510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58479-61-1
Molecular Formula C16H19ClSi
Molecular Weight (g/mol) 274.86
Synonym 1,1'-[Chloro(1,1-dimethylethyl)silylene]bisbenzene, Chloro(1,1-dimethylethyl)diphenylsilane, (1,1-Dimethylethyl)diphenylsilyl Chloride, (Chloro)(tert-butyl)diphenylsilane, Chloro(1,1-dimethylethyl)diphenylsilane, Chlorodiphenyl-tert-butylsilane, Diphenyl-tert-butylsilyl Chloride, NSC 617386, tert-Butylchlorodiphenylsilane, tert-Butyldiphenylsilyl Chloride, TBDPSCl, TBDPS-Cl
IUPAC Name tert-butyl-chloro-diphenylsilane
SMILES CC(C)(C)[Si](Cl)(c1ccccc1)c2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H19ClSi/c1-16(2,3)18(17,14-10-6-4-7-11-14)15-12-8-5-9-13-15/h4-13H,1-3H3
Formula Weight 274.0945
Chemical Name or Material tert-Butyldiphenylchlorosilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.